Learn More
TCR gamma/delta Monoclonal Antibody (5A6.E9), TRI-COLOR™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MHGD06
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
The TRI-COLOR® conjugate is a tandem R-phycoerythrin (PE)-Cy5® conjugate that permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry.Spezifikation
TCR gamma/delta | |
Monoclonal | |
TRI-COLOR | |
Trg | |
RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ | |
Mouse | |
purified | |
ASR | |
6964, 6965 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
5A6.E9 | |
PBS with 0.1% sodium azide | |
0 | |
TRD, Trg | |
Human TCR gamma/delta T cell receptor | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |