missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl tosylcarbamate, 95%, BLD Pharm™

Code produit 30801894
Encompass_Preferred
Modifier l'affichage
Click to view available options
Menge:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30801894 25 g
30801735 10 g
30801392 100 g
30791692 5 g
30789072 1 g
30795669 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30801894 Supplier BLD Pharm Supplier No. BD14170225g
 Eingeschränkter Artikel
Verfügbar ab: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material tert-Butyl tosylcarbamate
CAS 18303-04-3
Summenformel C12H17NO4S
MDL-Nummer MFCD00134267
Menge 25 g
SMILES O=C(OC(C)(C)C)NS(=O)(C1=CC=C(C)C=C1)=O
Molekulargewicht (g/mol) 271.33
Reinheit (%) 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.