missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrobiopterin, MedChemExpress
Description
Tetrahydrobiopterin ((Rac)-Sapropterin) is a cofactor of the aromatic amino acid hydroxylases enzymes and also acts as an essential cofactor for all nitric oxide synthase (NOS) isoforms.
Specifications
Specifications
| Chemischer Name oder Material | Tetrahydrobiopterin |
| Physikalische Form | Solid |
| CAS | 17528-72-2 |
| Farbe | White |
| Menge | 50 mg |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C9H15N5O3 |
| Zur Verwendung mit (Anwendung) | COVID-19-immunoregulation |
| Synonym | (Rac)-Sapropterin |
| Löslichkeitsinformationen | DMSO : 50 mg/mL (207.25 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?