Learn More
Invitrogen™ Human IL-1RA Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP10925
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Beschreibung
The RP-10925 protein is a recombinant protein suitable for use as a control in ELISA and Western blot applications. RP-10925 is reported to have the additional biological activity: ED50 as determined by the dose-dependent inhibition of IL-1 stimulation of D10S cells is 0.5ng/mL. Reconstitute with sterile,distilled water no less than 100ug/mL.
The RP-10925 protein is a recombinant protein suitable for use as a control in ELISA and Western blot applications. RP-10925 is reported to have the additional biological activity: ED50 as determined by the dose-dependent inhibition of IL-1 stimulation of D10S cells is 0.5ng/ml.Spezifikation
P18510 | |
protein with no preservative | |
Human IL1RA | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
IL-1ra | |
IL1RN | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
Control | |
3557 | |
20μg | |
N/A | |
RUO | |
Human IL1RA | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitroresearch only