Learn More
Invitrogen™ Human MMP-24 Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP77536
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Beschreibung
MMP24 Recombinant Protein, RP-77536, from Invitrogen.
RP-77536 is a purified recombinant MMP-24 (catalytic domain) protein purified from E. coli periplasm. MT5-MMP catalytic domain is produced by activation of a recombinant soluble proform of MT5-MMP. Activation generates two molecular species. One species begins at the N-terminus with Tyr156 of full-length MT5-MMP, while the other species is two amino acid residues shorter and starts with Leu158. Both species extend to Ser351 of MT5-MMP and they contain an additonal His-tag sequence at the C-terminal. The specific activity of MT5-MMP catalytic domain is greater than 120 mU/mg. 1 U is the activity that hydrolyzes 1μmol peptide (7-methoxycoumarin-4-yl) acetyl-Pro-Leu-Gly-Leu-(3-[2,4-dinitrophenyl]-L-2,3- diamino-propionyl)-Ala-Arg-NH2 (Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg) within 1 min. The catalytic domain of MT5-MMP is inhibited by tissue inhibitor of matrix metalloproteinase-2 (TIMP-2) and by chelators of divalent cations like EDTA or o-phenanthroline.Spezifikation
Q9Y5R2 | |
TBS with 5mM CaCl2 and no preservative; pH 7.5 | |
Human MMP24 | |
10μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
MT5-MMP | |
MMP24 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control | |
10893 | |
7.5 | |
E. coli | |
RUO | |
Human MMP24 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only