missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human S100B, Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ LFP0039
Weitere Details : Netto-Gewicht : 0.99000kg
Beschreibung
S100beta Recombinant Protein, LF-P0039, from Invitrogen.
S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer′s disease, Down′s syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes. This protein corresponds to human S100B (1-92 aa with N-GST tag), which has a molecular weight of 37 kDa. Purity: >90% by SDS-PAGE.Spezifikation
P04271 | |
Control | |
6285 | |
7.0 | |
E. coli | |
RUO | |
S100B | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
1mg/mL | |
0.02M HEPES with 10% glycerol and no preservative; pH 7.0 | |
Human S100B | |
500μg | |
-20°C or -80°C if preferred | |
Human S100B | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only.