missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-4 Rat anti-Mouse, Oligo, Clone: RMT4-54, BD™ AbSeq ™
Rat Monoclonal Antibody
Marke: BD Biosciences 940351
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
TIM-4 Monoclonal antibody specifically detects TIM-4 in Mouse samples. It is validated for Single Cell 3' SequencingSpezifikation
TIM-4 | |
Monoclonal | |
Oligo | |
TIMD-4, SMUCKLER, Tim4, Timd4 | |
Mouse Tim-4 Recombinant Protein | |
RUO | |
Mouse | |
IgG2a κ |
Single Cell 3' Sequencing | |
Clone RMT4-54 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |