missing translation for 'onlineSavingsMsg'
Learn More

Zinn(IV)-Isopropoxid, 99 % (Metallbasis), 10 % Gew./Vol. in Isopropanol, Thermo Scientific Chemicals

Catalog Number p-7059851
Click to view available options
Menge:
5 mL
25 mL
100 mL
500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1184-61-8
C12H32O4Sn
359.094
MFCD00145408
VUDAJANAMZGVOP-UHFFFAOYSA-N
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
85354423
Propan-2-ol;Zinn
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1184-61-8
C12H32O4Sn
359.094
MFCD00145408
VUDAJANAMZGVOP-UHFFFAOYSA-N
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
85354423
Propan-2-ol;Zinn
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]

Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Tin (IV) isopropoxide
Farbe Farblos
Geruch Alcohol-like
Hinweise zum Prozentgehalt (Metallbasis)
Strukturformel Sn[OCH(CH3)2]4
Menge 5 mL
UN-Nummer UN3274
Empfindlichkeit Moisture sensitive
Löslichkeitsinformationen Miscible with alcohols,ketones and esters.
Formelmasse 355.06
Reinheit (%) 99%
Physikalische Form Flüssig
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'