Learn More
Lovibond™ Tintometer™ Nitrite Nr. 1 Tabletten, Folienverpackung
Zur Verwendung mit Lovibond-Testkits, Kolorimetern und Spektralphotometern. Die Verwendung von Tablettenreagenzien garantiert eine genaue Reagenziendosierung und ermöglicht somit präzisere Titrationen als herkömmliche Tropfenzähler-Testkits
Marke: Lovibond™ 515200BT
Weitere Details : Netto-Gewicht : 0.07500kg
Beschreibung
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Spezifikation
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Nitrite No. 1 Tablets |