Learn More
Tirbanibulin, MedChemExpress
Product Code. 16474876
Encompass_Preferred
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 Milligramm
100 Milligramm
5 Milligramm
50 Milligramm
Product Code. 16474876
Brand: Medchem Express HY10340
om dit product te kopen Registreer vandaag om een webaccount aan te maken
MedChemExpress Tirbanibulin (KX2-391) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.
Tirbanibulin (KX2-391) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.
Specifications
Chemischer Name oder Material Tirbanibulin
CAS 897016-82-9
Farbe White
Hinweise zur Reinheitsqualität Research
Summenformel C26H29N3O3
Menge 100 mg
Synonym KX2-391 KX-01
Löslichkeitsinformationen DMSO : 41.67 mg/mL (96.56 mM; Need ultrasonic)
SMILES O=C(CC1=NC=C(C2=CC=C(OCCN3CCOCC3)C=C2)C=C1)NCC4=CC=CC=C4
Molekulargewicht (g/mol) 431.53
Formelmasse 431.53
Reinheit (%) 99.33%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease