Learn More
Eurofins DiscoverX TMDP Protein
Beschreibung
TMDP, also known as DUSP13 (Dual Specificity Phosphatase 13), is a member of the protein-tyrosine phosphatase superfamily cooperate with protein kinases to regulate cell proliferation and differentiation. This superfamily is separated into two families based on the substrate that is dephosphorylated. One family, the dual specificity phosphatases (DSPs) acts on both phosphotyrosine and phosphoserine/threonine residues. This gene encodes different but related DSP proteins through the use of non-overlapping open reading frames, alternate splicing, and presumed different transcription promoters. Expression of the distinct proteins from this gene has been found to be tissue specific and the proteins may be involved in postnatal development of specific tissues.
GenBank NM_016364
Spezifikation
Spezifikation
| Zusammensetzung | mg/mL of enzyme in 50 mM Tris/HCl pH 8.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekulargewicht | 49 kDa |
| Menge | 250 μg |
| Spezies | Human |
| Rekombinant | Recombinant |
| Proteinmarkierung | N-terminal GST |
| Expressionssystem | Recombinant enzyme expressed in E. coli cells |
| Proteinform | Full-length |
| Reinheits- oder Qualitätsgrad | ≥79% |
| Protein | TMDP, active |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.