missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Honeywell™
Click to view available options
Menge:
5 L
Verpackung:
Aluminum Can
Beschreibung
- ≥95% for general laboratory use.
Spezifikation
CAS | 108-88-3 |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
Reinheit (%) | min. 99.3 % |
Güte | Reagent |
Menge | 5 L |
Flammpunkt | 4 °C |
Verpackung | Aluminum Can |
Physikalische Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'