Learn More
Trazodone hydrochloride, Tocris Bioscience™
GREENER_CHOICE
5-HT2A and α1 adrenoceptor antagonist; also enhances neural differentiation; antidepressant and neuroprotectant
Marke:  Tocris Bioscience 6336/50
Encompass
Artikelnummer. 15746409
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Trazodone hydrochloride is a 5-HT2A and α1 adrenoceptor antagonist. Enhances neuronal differentiation of mouse and human neural progenitor cells. Also inhibits PERK/eIF2α-P-mediated reduction in protein synthesis and restores memory deficits in a mouse dementia model. Antidepressant and neuroprotectant.
Spezifikation
Spezifikation
2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one hydrochloride
25332-39-2
>99%
50 mg
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen