Learn More
TSPAN12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563538
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LMVPVQWSDM VTLKARMTNY GLPRYRWLTH AWNFFQREFK CCGVVYFTDW LEMTEMDWPP DSCCVREFPG CSKQAHQEDL SDLYQEGCGK KMY Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
TSPAN12 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.Spezifikation
TSPAN12 | |
Polyclonal | |
Unconjugated | |
TSPAN12 | |
9030619E17; AI426782; AI663988; AW111457; EVR5; MGC137611 protein; NET2; NET-2; Tetraspan NET-2; tetraspanin 12; tetraspanin-12; Tm4sf12; transmembrane 4 superfamily member 12; TSPAN12; Tspan-12; UNQ774/PRO1568; zgc:63880 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23554 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95859 | |
TSPAN12 | |
Recombinant protein corresponding to Human TSPAN12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only