missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550891
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total TSPAN13 / TM4SF13 protein.
TSPAN13 Polyclonal antibody specifically detects TSPAN13 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TSPAN13 | |
Polyclonal | |
Unconjugated | |
TSPAN13 | |
1100001I23Rik; NET6; NET-6; tetraspan NET-6; Tetraspan Tspan-13; tetraspanin 13; tetraspanin-13; Tm4sf13; transmembrane 4 superfamily member 13; transmembrane 4 superfamily member tetraspan NET-6; Tspan13; tspan-13; UNQ260/PRO296 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27075, 366602, 66109 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95857, Q5FVL6, Q9D8C2 | |
TSPAN13 | |
Synthetic peptide corresponding to a region derived from internal residues of human tetraspanin 13 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |