Learn More
TSPY10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561949
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EEEGLVERRE EAQRAQQAVP GPGPMTPESA LEELLAVQVE LEPVNAQARK AF Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 38%.
TSPY10 Polyclonal antibody specifically detects TSPY10 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TSPY10 | |
Polyclonal | |
Unconjugated | |
TSPY4 | |
testis specific protein, Y-linked 4; testis-specific Y-encoded protein 4; TSPY10; TSPY4; TSPY8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
728395 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0CV99 | |
TSPY4 | |
Recombinant protein corresponding to Human TSPY10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only