missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531842
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TTC14 is a protein coding gene.Spezifikation
TTC14 | |
Polyclonal | |
Unconjugated | |
TTC14 | |
2700016E08Rik; 4930434D01Rik; 4931403I22Rik; 4933402I15Rik; AI662468; AU014779; AW561908; cI-44; DRDL5813; Kiaa1980; mKIAA1980; PRO19630; tetratricopeptide repeat domain 14; tetratricopeptide repeat protein 14; TPR repeat protein 14; Ttc14; UNQ5813; UNQ5813/PRO19630 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96N46 | |
TTC14 | |
Recombinant protein fragment corresponding to a region within amino acids 470 and 664 of Human TTC14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |