Learn More
TTC17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558419
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DQPVRYHRGD IFENVDYVQF GEDSSTSSMM SVNFDVQSNQ SDINDSVKSS PVAHSILWIW GRDSDAYRDK QHILWPKRAD CTESYP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
TTC17 is a protein coding gene.Spezifikation
TTC17 | |
Polyclonal | |
Unconjugated | |
TTC17 | |
9130020K17Rik; D2Bwg1005e; tetratricopeptide repeat domain 17; tetratricopeptide repeat protein 17; Tetratricopeptide repeat protein 17-like protein; TPR repeat protein 17; Ttc17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55761 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AE7 | |
TTC17 | |
Recombinant protein corresponding to Human TTC17 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only