Learn More
TTLL10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560090
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KPTASNQGKG IFLLRNQEEV AALQAKTRSM EDDPIHHKTP FRGPQARVVQ RYIQNPLLVD GRKFDVRSYL LIACTTPYMI FFGHGYARLT LSLYDPHSSD LG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
Inactive polyglycylase.Spezifikation
TTLL10 | |
Polyclonal | |
Unconjugated | |
TTLL10 | |
4833412E22Rik; 4930595O22Rik; AU022799; BC042775; Inactive polyglycylase TTLL10; protein polyglycylase TTLL10; Ttll10; Ttll5; tubulin tyrosine ligase like 10; tubulin tyrosine ligase-like family member 10; tubulin tyrosine ligase-like family, member 10; tubulin tyrosine ligase-like family, member 5; tubulin--tyrosine ligase-like protein 10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254173 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZVT0 | |
TTLL10 | |
Recombinant protein corresponding to Human TTLL10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only