missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUB Recombinant Rabbit Monoclonal Antibody (24GB1795)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Spezifikation
| Antigen | TUB |
|---|---|
| Klon | 24GB1795 |
| Inhalt und Lagerung | -20°C |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30276038
![]() |
Invitrogen™
MA557297 |
100 μL |
500.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30275558
|
Invitrogen™
MA557298 |
20 μL |
146.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
TUB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the Tubby family of bipartite transcription factors. The encoded protein may play a role in obesity and sensorineural degradation. The crystal structure has been determined for a similar protein in mouse, and it functions as a membrane-bound transcription regulator that translocates to the nucleus in response to phosphoinositide hydrolysis. Two transcript variants encoding distinct isoforms have been identified for this gene.Spezifikation
| TUB | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| TUB | |
| CELE_F10B5.4; F10B5.4; Rd5; RDOB; retinal degeneration 5; TUB; TUB bipartite transcription factor; TUB-1; tubby bipartite transcription factor; tubby candidate gene; tubby homolog; tubby homologue; tubby protein; Tubby protein homolog; Tubby protein homolog 1 | |
| TUB | |
| Primary | |
| Affinity chromatography |
| 24GB1795 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88808, P50586, P50607 | |
| 22141, 25609, 7275 | |
| A synthesized peptide derived from human TUB1 (aa 1-50). | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts