Learn More
TUFM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527512
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (94%), Rat (95%), Rhesus Monkey (96%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein which participates in protein translation in mitochondria. Mutations in this gene have been associated with combined oxidative phosphorylation deficiency resulting in lactic acidosis and fatal encephalopathy. A pseudogene has been identified on chromosome 17.Spezifikation
TUFM | |
Polyclonal | |
Unconjugated | |
TUFM | |
2300002G02Rik; C76308; C76389; COXPD4; D250; EFTU; EF-Tu; EF-TuMT; elongation factor Tu; elongation factor Tu, mitochondrial; fi06f04; P43; Tu translation elongation factor, mitochondrial; TUFM; tufm {ECO:0000250; UniProtKB:P49411}; wu:fi06f04; zgc:110766 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7284 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
P49411 | |
TUFM | |
Recombinant fragment corresponding to a region within amino acids 189 and 377 of Human TUFM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |