missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541449
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MSQDNDTLMR DILGHELAAM RLQKLEQQRR LFEKKQRQKR QELLMVQANP Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Rat: 86%
TULP2 Polyclonal antibody specifically detects TULP2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
TULP2 | |
Polyclonal | |
Unconjugated | |
TULP2 | |
cancer testis antigen 65; cancer/testis antigen 65; CT65; Pdet; Protein P4-6; TUB like protein 2; tubby like protein 2; tubby-like protein 2; Tubby-related protein 2; TUBL2; TULP2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7288 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00295 | |
TULP2 | |
Synthetic peptide directed towards the N-terminal of human TULP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |