Learn More
TUSC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562408
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TNRRARDSGR EDEPGSPRAL RARLEKLEAM YRRALLQLHL EQRGPRPSGD KEEQPLQEPD SGLRSRDSEP S Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 33%.
This gene is located within the region of chromosome 9p that harbors tumor suppressor genes critical in carcinogenesis. It is an intronless gene which is downregulated in non-small-cell lung cancer and small-cell lung cancer cell lines, suggesting that it may play a role in lung tumorigenesis.Spezifikation
TUSC1 | |
Polyclonal | |
Unconjugated | |
TUSC1 | |
2200001D17Rik; TSG9; TSG-9; tumor suppressor candidate 1; tumor suppressor candidate gene 1 protein; Tumor suppressor candidate gene 1 protein homolog; tumour suppressor candidate 1; TUSC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286319 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2TAM9 | |
TUSC1 | |
Recombinant protein corresponding to Human TUSC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only