Learn More
DuPont™ Tychem™ 6000 F Plus with Attached Gloves Model CHZ5
Artikelnummer. 30305896
Change view
Click to view available options
Größe:
X-Small
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
Packungsgröße:
10 Stück
Artikelnummer. Größe unitSize
30305896 Small 10 Stück
30305933 Medium 10 Stück
30305968 Large 10 Stück
30305953 X-Small 10 Stück
30305966 2X-Large 10 Stück
30305960 3X-Large 10 Stück
30305945 4X-Large 10 Stück
30305938 5X-Large 10 Stück
8 options
Artikelnummer. 30305896
Marke: DuPont™ D15546500
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Hooded coverall with attached gloves. Stitched and over-taped seams. Double cuffs with attached chemical barrier gloves on inner cuff.
  • DuPont™ Tychem™ 6000 F Plus with attached non-dissipative undergloves, TFCHZ5TGY08. Hooded coverall. Stitched and over-taped seams. Double cuffs with attached chemical barrier gloves on inner cuff. Elastication at wrists, ankles, face, and waist. Double zippers and double flaps. Outer flap with adhesive tape. Grey.
  • Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek ™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
  • Tychem™ 6000 F garments are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Hooded Coveralls
Größe Small
Geschlecht Unisex
Material HDPE
Farbe Gray
Einwegprodukt Single-use
Taillenform Elastic
Knöchelform Elastic
Stulpenform Elastic
Flaschenhalstyp Hooded
Hood Type Elastic
Verschlusstyp Double Zipper Front with Double Flaps
Saumtyp Stitched, Taped
Sterilität Non-sterile
Widerstand Liquid