missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F Plus with Attached Gloves Model CHZ5
Click to view available options
Größe:
X-Small
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
Packungsgröße:
10 Stück
Beschreibung
- DuPont™ Tychem™ 6000 F Plus with attached non-dissipative undergloves, TFCHZ5TGY08. Hooded coverall. Stitched and over-taped seams. Double cuffs with attached chemical barrier gloves on inner cuff. Elastication at wrists, ankles, face, and waist. Double zippers and double flaps. Outer flap with adhesive tape. Grey.
- Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek ™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
- Tychem™ 6000 F garments are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Spezifikation
Spezifikation
| Produkttyp | Hooded Coveralls |
| Größe | X-Small |
| Geschlecht | Unisex |
| Material | HDPE |
| Farbe | Gray |
| Einwegprodukt | Single-use |
| Taillenform | Elastic |
| Knöchelform | Elastic |
| Stulpenform | Elastic |
| Flaschenhalstyp | Hooded |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur