missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F Plus with Dissipative Socks and Attached Gloves Model CHZ5
Description
- DuPont ™ Tychem™ 6000 F Plus with attached non-dissipative undergloves and dissipative socks, TFCHZ5TGY26. Hooded coverall. Stitched and over-taped seams. Double cuffs with attached non-dissipative undergloves on inner cuff. Elastication at wrists,ankles, face, and waist. Double zippers and double flaps. Outer flap with adhesive tape. Integrated dissipative socks. Gray.
- Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
- Tychem™ 6000 F garments are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Specifications
Specifications
| Produkttyp | Hooded Coveralls with Socks |
| Größe | Medium |
| Geschlecht | Unisex |
| Material | HDPE |
| Farbe | Gray |
| Einwegprodukt | Single-use |
| Taillenform | Elastic |
| Knöchelform | Elastic |
| Stulpenform | Double |
| Flaschenhalstyp | Hooded |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?