Learn More
Tyrosine Hydroxylase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen P21962
Weitere Details : Netto-Gewicht : 0.00450kg
Beschreibung
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
Tyrosine Hydroxylase | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P04177, P07101 | |
TH | |
SDS-denatured rat tyrosine hydroxylase, purified from pheochromocytoma. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
TH | |
dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase | |
Rabbit | |
purified | |
RUO | |
25085, 7054 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |