Learn More
DuPont™ Tyvek™ 400 DualCombi Model TD0125S
Artikelnummer. 30305905
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packungsgröße:
25 Stück
Artikelnummer. 30305905
Marke: DuPont™ D14681398
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Collared coverall with Tyvek at the front and large breathable polypropylene back panel. Stitched external seams in red. Elasticated wrists and ankles.
  • DuPont™ Tyvek™ 400 DualCombi. Collared coverall combining Tyvek™ with a spunbound polypropylene back panel. Available in sizes SM to 4X. Elasticated wrists, waist and ankles. Stitched external seams in red. Thumb hole on sleeve. White.
  • The Tyvek™ front provides an ideal balance of protection, durability and comfort. It is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size. It is ultra-low-linting and antistatically treated. Silicon non-added.
  • The large back panel from shoulders to ankle is made of spunbound polypropylene: it is breathable and offers high level of comfort but no protection.
  • Tyvek™ 400 DualCombi is designed for work environment where comfort is important and exposure risk is limited to the front. Typical application is the handling of composite materials, paint spraying, glass manufacturing and utilities.
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Collared Coveralls
Größe Large
Geschlecht Unisex
Material Polyethylene
Farbe White
Einwegprodukt Single-use
Taillenform Elastic (Stitched-in)
Knöchelform Elastic
Stulpenform Elastic
Flaschenhalstyp Collared
Hood Type Unhooded
Verschlusstyp Front Zipper with Flap
Saumtyp Stitched
Sterilität Non-sterile
Widerstand Liquid