Learn More
DuPont™ Tyvek™ 400 DualFinish Model TD0127S
Artikelnummer. 30305907
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packungsgröße:
25 Stück
Artikelnummer. 30305907
Marke: DuPont™ D14681302
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Hooded coverall with Tyvek at the front and large breathable SMS back. Stitched external seams in blue. Elasticated wrists, ankles and face.
  • DuPont™ Tyvek™ 400 DualFinish. Hooded coverall combining Tyvek™ with a SMS back panel from shoulders to ankle. Available in sizes SM to 4X. 2-piece hood. Elasticated face, wrists, waist and ankles. Stitched external seams in blue. Thumb hole on sleeve. White.
  • The Tyvek™ front provides an ideal balance of protection, durability and comfort. It is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size. It is ultra-low-linting and antistatically treated. Silicon non-added.
  • The large, breathable back panel, made of SMS nonwoven, offers lower protection against particles and light water-based splashes, yet high levels of comfort.
  • Tyvek™ 400 DualFinish is designed for specific applications that demand comfort while helping to provide protection from frontal exposure during paint spraying or any work involving composite materials.
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Hooded Coveralls
Größe Small
Geschlecht Unisex
Material Polyethylene
Farbe White
Einwegprodukt Single-use
Taillenform Standard
Knöchelform Elastic
Stulpenform Elastic
Flaschenhalstyp Hooded
Hood Type Elastic
Verschlusstyp Zipper
Saumtyp Stitched
Sterilität Non-sterile
Widerstand Liquid