missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ TYVEK™ 500 HP Model TY178
Click to view available options
Größe:
X-Small
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
Packungsgröße:
25 Stück
Beschreibung
- Workers operating with full-body harness in environments where hazardous particle or light liquid aerosol hazards are present, previously had to compromise between efficient personal protection and protecting their full-body harness against external contamination.
- The new Tyvek™ 500 HP TY178S HP garment offers a unique solution - protection of both the wearer and the full-body harness - by providing a barrier between the user and the particle or light liquid aerosol hazards in the environment. Designed to be worn over a full-body harness, the innovative, patent-pending garment design allows workers to access and utilize multiple D-Rings on their full-body harness while providing an ideal balance of protection, durability, and comfort.
- This new TY178S HP model has four attached sleeves for use with front, back and side D-Rings of a full-body harness. The off-centered zipper entry allows accommodating the front D-Ring sleeve for front D-Ring connection. Compatible with variety of full-body harness models, this garment may be used for a wide range of applications including painting, cleaning, maintenance, and more.
- In addition to its innovative design, this garment also offers an ideal balance of protection and durability thanks to the unique Tyvek™ fabric. It is also extremely lightweight offering high level of comfort and mobility. The garment is low linting (suitable also for paint applications). Silicone non-added.
Spezifikation
Spezifikation
| Produkttyp | Collared Coveralls |
| Größe | 3X-Large |
| Geschlecht | Unisex |
| Material | HDPE |
| Farbe | White |
| Einwegprodukt | Single-use |
| Taillenform | Elastic |
| Knöchelform | Elastic |
| Stulpenform | Elastic |
| Flaschenhalstyp | Hooded |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur