Learn More
DuPont™ Tyvek™ 500 TY198S AV
Artikelnummer. p-200112405
Click to view available options
Größe:
X-Small
Small
Medium
Large
2X-Large
3X-Large
Packungsgröße:
25 Stück
Artikelnummer. 30305894
Marke: DuPont™ D15570488
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Hooded coverall with ergonomic-protective design. Stitched external seams. Elasticated wrists, ankles and face.
  • DuPont™ Tyvek™ 500 AV. Hooded coverall available in white, in sizes SM to 3X. Robust yet lightweight. 3-piece hood for optimal fit to head and face when turning. Elasticated face, wrists and ankles as well as glued-in waist elastic. Ample crotch area for freedom of movement. Large, easy-to-grasp zipper puller.
  • Tyvek™ garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek™ is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated on both sides. Silicone non-added.
  • Applications of Tyvek™ 500 AV garments include routine tasks in low-risk situations, or when risk is defined, in context of BSL3 or higher, and low exposure to body fluids: transport of infected patient, patient with severe conditions, hospital setting.
TRUSTED_SUSTAINABILITY
Spezifikation
Größe Small
Geschlecht Unisex
Material HDPE
Farbe White
Einwegprodukt Single-use
Taillenform Elastic
Knöchelform Elastic
Stulpenform Elastic
Flaschenhalstyp Hooded
Hood Type Elastic
Verschlusstyp Front Zipper with Flap
Saumtyp Stitched
Sterilität Non-sterile
Widerstand Liquid