Learn More
DuPont™ Tyvek™ 500 TY198S ES
Artikelnummer. 30305903
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
25 Stück
Artikelnummer. 30305903
Marke: DuPont™ D15570298
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Hooded coverall with ergonomic-protective design. Stitched external seams. Elasticated wrists, ankles and face.
  • DuPont™ Tyvek™ 500 ES. Hooded coverall available in white, in sizes SM to 3X. Robust yet lightweight. 3-piece hood for optimal fit to head and face when turning. Elasticated face, wrists and ankles as well as glued-in waist elastic. Ample crotch area for freedom of movement. Large, easy-to-grasp zipper puller.
  • Tyvek™ garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek™ is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated on the inside. Silicone non-added.
  • Applications of Tyvek™ 500 ES garments include routine tasks in low-risk situations, or when risk is defined, in context of BSL3 or less, and low exposure to body fluids: transport of infected patients, testing, patient triage, refugee camp preparation, people rescue.
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Hooded Coveralls
Größe 2X-Large
Geschlecht Unisex
Material HDPE
Farbe White
Einwegprodukt Single-use
Taillenform Elastic
Knöchelform Elastic
Stulpenform Elastic
Flaschenhalstyp Hooded
Hood Type Elastic
Verschlusstyp Front Zipper with Flap
Saumtyp Stitched
Sterilität Non-sterile
Widerstand Liquid