Learn More
DuPont™ Tyvek™ 800 TJ198T ES
Artikelnummer. p-200112406
Click to view available options
Größe:
X-Small
Small
Medium
Large
2X-Large
3X-Large
Packungsgröße:
25 Stück
Artikelnummer. 30305936
Marke: DuPont™ D15570650
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Hooded coverall with stitched and over-taped seams. Thumb loops. Elastication at wrists, ankles and face.
  • DuPont™ Tyvek™ 800 ES. Hooded coverall available in sizes SM to 3X. White. Robust yet lightweight. Self-adhesive zipper flap. Self-adhesive chin flap for tight seal of suit to mask. Elasticated face, wrists and ankles as well as glued-in waist elastic. Thumb loops to prevent sleeves from riding up.
  • Tyvek™ 800 ES garments are composed of PE laminated nonwoven, providing an ideal balance of protection, durability and comfort. They benefit from the use of a specific 'impervious technology' to achieve a high protection against biological hazards (EN 14126) including virus protection with an improved breathability. The fabric is antistatically treated.
  • Applications for Tyvek™ 800 ES garments include delivering health care in high-risk situations, or where risk is unknown, in context of BSL3 or higher (Tuberculosis, Covid) with high exposure to body fluids: urgent medical help, road accidents, catastrophes.
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Hooded Coveralls
Größe Small
Geschlecht Unisex
Material HDPE
Farbe White
Einwegprodukt Single-use
Taillenform Elastic
Knöchelform Elastic
Stulpenform Elastic
Flaschenhalstyp Hooded
Hood Type Elastic
Verschlusstyp Zipper
Saumtyp Stitched, Taped
Sterilität Non-sterile
Widerstand Liquid