Learn More
Umbralisib, MedChemExpress
Artikelnummer. 16467777
Encompass_Preferred
Change view
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Packungsgröße:
5 Milligramm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16467777 5 mg 5 Milligramm
16445796 100 mg -
16435058 10 mg -
16411297 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16467777 Lieferant Medchem Express Lieferanten-Nr. HY122795mg
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
MedChemExpress Umbralisib (TGR-1202) is an orally active, potent and selective dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibitor, with EC50 of 22.2 nM and 6.0 μM, respectively. Umbralisib exhibits unique immunomodulatory effects on chronic lymphocytic leukemia (CLL) T cells. Umbralisib can be used for haematological malignancies reseach.
Umbralisib (TGR-1202) is an orally active, potent and selective dual PI3Kδ and casein kinase-1-ε (CK1ε) inhibitor, with EC50 of 22.2 nM and 6.0 μM, respectively. Umbralisib exhibits unique immunomodulatory effects on chronic lymphocytic leukemia (CLL) T cells. Umbralisib can be used for haematological malignancies reseach.
Spezifikation
Chemischer Name oder Material Umbralisib
CAS 1532533-67-7
Farbe White
Hinweise zur Reinheitsqualität Research
Summenformel C31H24F3N5O3
Menge 5 mg
Synonym TGR-1202 RP5264
Löslichkeitsinformationen DMSO : 100 mg/mL (174.96 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=CC(F)=C2)=C([C@@H](N3N=C(C4=CC=C(OC(C)C)C(F)=C4)C5=C(N)N=CN=C53)C)OC6=CC=C(F)C=C16
Molekulargewicht (g/mol) 571.55
Formelmasse 571.55
Reinheit (%) 95.05%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease