Learn More
DiscoverX Uridine 5'-Triphosphate
Click to view available options
Menge:
10 mg
Beschreibung
Uridine 5'-Triphosphate (UTP) is a uracil nucleoside triphosphate that acts as a substrate for RNA synthesis during transcription. UTP also provides a source of energy and activates substrates in metabolic reactions, like ATP.
Spezifikation
Spezifikation
| Produkttyp | Control Ligands |
| Menge | 10 mg |