missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ USP48 (Human) Recombinant Protein
Human USP48 full-length ORF ( NP_115612.4, 1 a.a. - 1035 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00084196-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MAPRLQLEKAAWRWAETVRPEEVSQEHIETAYRIWLEPCIRGVCRRNCKGNPNCLVGIGEHIWLGEIDENSFHNIDDPNCERRKKNSFVGLTNLGATCYVNTFLQVWFLNLELRQALYLCPSTCSDYMLGDGIQEEKDYEPQTICEHLQYLFALLQNSNRRYIDPSGFVKALGLDTGQQQDAQEFSKLFMSLLEDTLSKQKNPDVRNIVQQQFCGEYAYVTVCNQCGRESKLLSKFYELELNIQGHKQLTDCISEFLKEEKLEGDNRYFCENCQSKQNATRKIRLLSLPCTLNLQLMRFVFDRQTGHKKKLNTYIGFSEILDMEPYVEHKGGSYVYELSAVLIHRGVSAYSGHYIAHVKDPQSGEWYKFNDEDIEKMEGKKLQLGIEEDLAEPSKSQTRKPKCGKGTHCSRNAYMLVYRLQTQEKPNTTVQVPAFLQELVDRDNSKFEEWCIEMAEMRKQSVDKGKAKHEEVKELYQRLPAGAEPYEFVSLEWLQKWLDESTPTKPIDNHACLCSHDKLHPDKISIMKRISEYAADIFYSRYGGGPRLTVKALCKECVVERCRILRLKNQLNEDYKTVNNLLKAAVKGSDGFWVGKSSLRSWRQLALEQLDEQDGDAEQSNGKMNGSTLNKDESKEERKEEEELNFNEDILCPHGELCISENERRLVSKEAWSKLQQYFPKAPEFPSYKECCSQCKILEREGEENEALHKMIANEQKTSLPNLFQDKNRPCLSNWPEDTDVLYIVSQFFVEEWRKFVRKPTRCSPVSSVGNSALLCPHGGLMFTFASMTKEDSKLIALIWPSEWQMIQKLFVVDHVIKITRIEVGDVNPSETQYISEPKLCPECREGLLCQQQRDLREYTQATIYVHKVVDNKKVMKDSAPELNVSSSETEEDKEEAKPDGEKDPDFNQSNGGTKRQKISHQNYIAYQKQVIRRSMRHRKVRGEKALLVSANQTLKELKIQIMHAFSVAPFDQNLSIDGKILSDDCATLGTLGVIPESVILLKADEPIADYAAMDDVMQVCMPEEGFKGTGLLGH
Spezifikation
NP_115612.4 | |
ubiquitin specific peptidase 48 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
USP48 | |
GST |
84196 | |
Wheat germ expression system | |
25 μg | |
DKFZp762M1713, MGC132556, MGC14879, RAP1GA1, USP31 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |