missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BD Vacutainer Serumkanülen 5ml VE=100 Stück
Marke: BD BP367614
Weitere Details : Netto-Gewicht : 0.10000kg
Spezifikation
Serum Tube | |
Hemogard | |
13 x 75 mm |
Glass | |
5 mL | |
Used to obtain a serum sample |