missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP3/Cellubrevin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10702-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Because of its high homology to other known VAMPs, its broad tissue distribution, and its subcellular localization, the protein encoded by this gene was shown to be the human equivalent of the rodent cellubrevin. In platelets the protein resides on a compartment that is not mobilized to the plasma membrane on calcium or thrombin stimulation.Spezifikation
VAMP3/Cellubrevin | |
Polyclonal | |
Unconjugated | |
VAMP3 | |
CEB, Cellubrevin, SYB3, Synaptobrevin 3, VAMP 3, VAMP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22319, 29528, 9341 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63024, P63025, Q15836 | |
VAMP3 | |
VAMP3/Cellubrevin Fusion Protein Ag1156 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |