missing translation for 'onlineSavingsMsg'
Learn More

Varespladib, MedChemExpress

Artikelnummer. 16415668
Encompass_Preferred
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 Milligramm
50 Milligramm
This item is not returnable. View return policy

Product Code. 16415668

Brand: Medchem Express HY13402

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress Varespladib (LY315920) is a potent and selective group IIA, secretory phospholipase A2 (sPLA2) inhibitor with an IC50 of 9 nM. Varespladib exhibits the significant inhibitory effect on sPLA2 activity in serum from various species including rat, rabbit, guinea pig and human with IC50s of 8.1 nM, 5.0 nM, 3.2 nM and 6.2 nM, respectively.

Varespladib (LY315920) is a potent and selective group IIA, secretory phospholipase A2 (sPLA2) inhibitor with an IC50 of 9 nM. Varespladib exhibits the significant inhibitory effect on sPLA2 activity in serum from various species including rat, rabbit, guinea pig and human with IC50s of 8.1 nM, 5.0 nM, 3.2 nM and 6.2 nM, respectively.

Specifications

Chemischer Name oder Material Varespladib
CAS 172732-68-2
Farbe Off-White
Hinweise zur Reinheitsqualität Research
Summenformel C21H20N2O5
Menge 10 mg
Synonym LY315920
Löslichkeitsinformationen DMSO : ≥ 32 mg/mL (84.12 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)COC1=CC=CC2=C1C(C(C(N)=O)=O)=C(CC)N2CC3=CC=CC=C3
Molekulargewicht (g/mol) 380.39
Formelmasse 380.39
Reinheit (%) 98.68%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.