Learn More
Nabertherm VCD Software
For Visualization, Control, and Documentation. VCD Software is used to record process data from the controllers B400/B410, C440/C450 and P470/P480. Up to 400 different heat treatment programs can be stored.
Marke:  Nabertherm 635004763
Weitere Details : Netto-Gewicht : 0.20000kg
Artikelnummer. 15774027
  • 1180.00 € / 1 Stück
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Available for controllers B400/B410/C440/C450/P470/P480
  • Suitable for operating systems Microsoft Windows 7 (32/64 Bit) or 8/8.1 (32/64 Bit)
  • Simple installation
  • Setting, Archiving and print of programs and graphics
  • Operation of controllers via PC
  • Archiving of process curves from up to 16 furnaces (also multi-zone controlled)
  • Redundant saving of archives on a server drive
  • Higher security level due to binary data storage
  • Free input of charge date with comfortable search function
  • Possibility to evaluate data, files can be converted to Excel
  • Generation of a PDF-report
  • Language selection: German, English, Italian, French, Spanish, Russian
Spezifikation
Spezifikation
Software
For Visualization, Control, and Documentation.