missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VEGF-B186 Antibody, R&D Systems™
Rat Monoclonal Antibody
558.30 €
Spezifikation
Antigen | VEGF-B |
---|---|
Klon | 124112 |
Verdünnung | Western Blot 1 ug/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18182451
|
R&D Systems
MAB767 |
500 μg |
558.30 €
500 Mikrogramm |
Verfügbar ab: 31-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot.Spezifikation
VEGF-B | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL | |
Vegfa | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
124112 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P49766.2 | |
7423 | |
E. coli-derived recombinant mouse VEGF-B 186 Pro22-Ala207 Accession # P49766.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse VEGF-B 186 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) VEGF 165 rhVEGF-B 186 recombinant mouse (rm) VEGF-B 167 rhVEGF-C, rmVEGF-D, rhCTGF, rhPlGF, or rhLDGF is observed. |
For Research Use Only