Learn More
VIAAT/SLC32A1/VGAT Antibody (CL2793), Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-46628-25ul
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 18662316
  • 369.00 € / 25 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
VIAAT/SLC32A1/VGAT Monoclonal antibody specifically detects VIAAT/SLC32A1/VGAT in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
VIAAT/SLC32A1/VGAT
Monoclonal
Unconjugated
PBS (pH 7.2), 40% Glycerol
bA122O1.1, hVIAAT, solute carrier family 32 (GABA vesicular transporter), member 1, Solute carrier family 32 member 1, Vesicular GABA transporter, vesicular inhibitory amino acid transporter, VGATGABA and glycine transporter, VIAAT
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence ATSVSNKSQAKMSGMFARMGFQAATDEEAVGFAHCDDLDFEHRQGLQMDILKAEGEPCGDEGAEAPVEGDIHYQRGSGAPLPPSGSKDQVG
25 μL
Neuroscience, Neurotransmission
140679
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL2793
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Q9H598
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat