missing translation for 'onlineSavingsMsg'
Learn More

Vitamin B12, Serva Electrophoresis™

Product Code. 11707896
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
11707896 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11707896 missing translation for 'mfr' Serva Electrophoresis missing translation for 'supplierNo' 3831002
 Eingeschränkter Artikel
Verfügbar ab: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 68-19-9
Summenformel C63H89CoN14O14P
Menge 500 mg
Synonym vitamin b12, cyanocobalamin, cobalamin, cyanocobalamin;cyanocob iii alamin, crystamine, anacobin, berubigen, cobavite, depinar, nascobal
InChI-Schlüssel AGVAZMGAQJOSFJ-UHFFFAOYNA-M
Molekulargewicht (g/mol) 1356.40
PubChem CID 134691994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.