Learn More
von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody.
Marke: Epredia MS-690-P
Beschreibung
Molecular genetic studies suggest that VHL is a tumor-suppressor gene. Alterations in the von Hippel-Lindau (VHL) gene are correlated with a diverse group of neoplasms including hemangioblastoma, clear cell renal carcinoma (RCC), and pheochromocytoma. Two transcriptional elongation factors, Elongin B and C, are shown to bind in vitro and in vivo to a short, colinear region of the VHL protein (pVHL) that is frequently mutated in human tumors. It is proposed that pVHL functions in vivo as a negative regulator of transcription elongation.Host Species: Mouse
Clone: Ig33
Isotype: IgG1/κ
Species Reactivity: Human and Mouse. Others not known.
Epitope: aa 1-54
Immunogen: A recombinant protein containing aa 1-213 of human vHL
Molecular Weight: 30kDa
Positive Control: Kidney
Cellular Localization: Cytoplasmic Ab-1 does not recognize the truncated form of VHL protein.
Note:
USA: RUO; Int'l: RUO
Spezifikation
| von Hippel-Lindau (vHL) Protein Ab-1 | |
| Ig33 | |
| Unconjugated | |
| A recombinant protein containing aa 1-213 of human VHL | |
| RUO | |
| Primary | |
| Concentrated |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 1 mL | |
| Cancer and Tumor Biology | |
| Human, Mouse | |
| IgG2a κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.