Learn More
von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody.
Marke:  Epredia MS-690-PABX
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 12692037
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Molecular genetic studies suggest that VHL is a tumor-suppressor gene. Alterations in the von Hippel-Lindau (VHL) gene are correlated with a diverse group of neoplasms including hemangioblastoma, clear cell renal carcinoma (RCC), and pheochromocytoma. Two transcriptional elongation factors, Elongin B and C, are shown to bind in vitro and in vivo to a short, colinear region of the VHL protein (pVHL) that is frequently mutated in human tumors. It is proposed that pVHL functions in vivo as a negative regulator of transcription elongation.
Host Species: Mouse
Clone: Ig33
Isotype: IgG1/κ
Species Reactivity: Human and Mouse. Others not known.
Epitope: aa 1-54
Immunogen: A recombinant protein containing aa 1-213 of human vHL
Molecular Weight: 30kDa
Positive Control: Kidney
Cellular Localization: Cytoplasmic Ab-1 does not recognize the truncated form of VHL protein.
Note:
USA: RUO; Int'l: RUO
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
von Hippel-Lindau (vHL) Protein Ab-1
Ig33
Unconjugated
A recombinant protein containing aa 1-213 of human VHL
RUO
Primary
Concentrated
Monoclonal
1mg/mL
Mouse
200 μL
Cancer and Tumor Biology
Human, Mouse
IgG2a κ