Learn More
Invitrogen™ VPS18 Recombinant Rabbit Monoclonal Antibody (24GB2405)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557319
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
VPS18 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
VPS18 is a vesicle mediated protein that plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Spezifikation
| VPS18 | |
| Recombinant Monoclonal | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R307, Q9P253 | |
| VPS18 | |
| A synthesized peptide derived from human VPS18 (aa 1-28). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2405 | |
| Unconjugated | |
| VPS18 | |
| 9930024E13Rik; hVPS18; KIAA1475; PEP3; vacuolar protein sorting 18 (yeast); vacuolar protein sorting 18 homolog; vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting protein 18; vacuolar protein sorting-associated protein 18 homolog; Vps18; VPS18 CORVET/HOPS core subunit; VPS18, CORVET/HOPS core subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 228545, 296083, 57617 | |
| -20°C | |
| Liquid |