Learn More
VSIG4 Monoclonal Antibody (NLA14), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
176.00 € - 446.00 €
Spezifikation
Antigen | VSIG4 |
---|---|
Klon | NLA14 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16311504
|
Affymetrix eBioscience
63-5752-80 |
25 μg |
176.00 €
25 Mikrogramm |
Verfügbar ab: 07-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16321504
|
Affymetrix eBioscience
63-5752-82 |
100 μg |
446.00 €
100 Mikrogramm |
Verfügbar ab: 07-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: This NLA14 monoclonal antibody recognizes mouse V-Set and Immunoglobulin domain containing 4 (VSIG4), also known as Complement Receptor of the Immunoglobulin superfamily (CRIg) or Z39Ig. VSIG4 is a type I transmembrane glycoprotein structurally related to the B7 family of immune regulatory proteins. It contains one complete V-type Ig domain and one truncated C-type Ig domain. VSIG4 is exclusively expressed on tissue resident and tumor infiltrating macrophages. It has been shown to bind complement components C3b and iC3b. This binding inhibits the alternative complement pathway and facilitates phagocytosis of complement-opsonized pathogens. VSIG4 has also been reported to suppress T cell activation, proliferation and IL-2 production thereby playing a role in the maintece of peripheral T cell tolerance and suppression of established inflammation. Expression of VSIG4 on tumor-infiltrating macrophages suggests its role in immune evasion. Pro-inflammatory stimuli such as TNF and LPS have been reported to down-regulate the expression of VSIG4. Peritoneal macrophages in Balb/c mice express significantly higher levels of VSIG4 than such macrophages in C57Bl/6 or Swiss Webster mice. The NLA14 antibody will recognize VSIG4 on cells that have been formaldehyde-fixed and permeabilized. This antibody does not block the ligation of VSIG4 to its T cell ligand. The NLA14 antibody does not cross-react with rat or human VSIG4.
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.Spezifikation
VSIG4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide, pH 7.2 | |
278180 | |
Fc protein and C6 cells | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
VSIG4 |
NLA14 | |
Flow Cytometry | |
Super Bright 600 | |
Rat | |
Mouse | |
A530061A11, BC025105, complement receptor of the immunoglobulin superfamily, CRIg, Ig superfamily protein, MGC137243 protein, protein Z39Ig, UNQ317/PRO362, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, VSIG4, Z39IG | |
VSIG4 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |