Learn More
WAPL/WAPAL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16370-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Transcriptional repressor which may play a role in development of the central nervous system (CNS).Spezifikation
WAPL/WAPAL | |
Polyclonal | |
Unconjugated | |
WAPL | |
FOE, Friend of EBNA2 protein, KIAA0261, WAPAL, WAPL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
218914, 23063 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q65Z40, Q7Z5K2 | |
Wapl | |
WAPL; WAPAL Fusion Protein Ag9280 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |