Learn More
Merck Chemicals Water standard 1% Standard for volumetric Karl Fischer Titration 1 g «» 10 mg H2O apura®
Marke:  Merck Chemicals 1880520010
Encompass_Preferred
Water standard 1% Standard for volumetric Karl Fischer Titration 1 g «» 10 mg H2O apura®
Artikelnummer. 15382892
  • 136.73 € / 1 Packung