missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR38 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25683-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
WDR38 | |
Polyclonal | |
Unconjugated | |
WDR38 | |
WD repeat domain 38, WDR38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
401551 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5JTN6 | |
WDR38 | |
WDR38 Fusion Protein Ag22444 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |